missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dibromopyrazine 98.0+%, TCI America™
$44.73 - $128.62
Chemical Identifiers
| CAS | 23229-26-7 |
|---|---|
| Molecular Formula | C4H2Br2N2 |
| Molecular Weight (g/mol) | 237.88 |
| MDL Number | MFCD08061601 |
| InChI Key | KIYKHEOWZLJZSB-UHFFFAOYSA-N |
| PubChem CID | 11470421 |
| IUPAC Name | 2,5-dibromopyrazine |
| SMILES | BrC1=CN=C(Br)C=N1 |
Chemical Identifiers
| 23229-26-7 | |
| 237.88 | |
| KIYKHEOWZLJZSB-UHFFFAOYSA-N | |
| 2,5-dibromopyrazine |
| C4H2Br2N2 | |
| MFCD08061601 | |
| 11470421 | |
| BrC1=CN=C(Br)C=N1 |
Specifications
| 23229-26-7 | |
| White-Yellow | |
| MFCD08061601 | |
| KIYKHEOWZLJZSB-UHFFFAOYSA-N | |
| 2,5-dibromopyrazine | |
| 11470421 | |
| ≥98.0% (GC) | |
| 2,5-Dibromopyrazine |
| 47°C | |
| C4H2Br2N2 | |
| 1 g | |
| BrC1=CN=C(Br)C=N1 | |
| 237.88 | |
| 237.88 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator