accessibility menu, dialog, popup

UserName

NNBrBr

CAS RN 23229-26-7

IUPAC Name: 2,5-dibromopyrazine
Synonyms: 2,5-dibromopyrazine
Molecular Weight (g/mol): 237.88
Molecular Formula: C4H2Br2N2
InChi Key: KIYKHEOWZLJZSB-UHFFFAOYSA-N
SMILES: BrC1=CN=C(Br)C=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results