missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromopyrazine 98.0+%, TCI America™

Catalog No. D43231G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43231G 1 g
D43235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43231G Supplier TCI America Supplier No. D43231G
Only null left

Chemical Identifiers

CAS 23229-26-7
Molecular Formula C4H2Br2N2
Molecular Weight (g/mol) 237.88
MDL Number MFCD08061601
InChI Key KIYKHEOWZLJZSB-UHFFFAOYSA-N
PubChem CID 11470421
IUPAC Name 2,5-dibromopyrazine
SMILES BrC1=CN=C(Br)C=N1

Specifications

Melting Point 47°C
Color White-Yellow
Quantity 1 g
Formula Weight 237.88
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromopyrazine
TSCA No
Recommended Storage Refrigerator