missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dibromofuran (stabilized with MgO) 97.0+%, TCI America™
$618.87 - $2111.97
Chemical Identifiers
| CAS | 32460-00-7 |
|---|---|
| Molecular Formula | C4H2Br2O |
| Molecular Weight (g/mol) | 225.87 |
| MDL Number | MFCD00191342 |
| InChI Key | LACYYWKMIJOHLU-UHFFFAOYSA-N |
| PubChem CID | 10878846 |
| IUPAC Name | 2,5-dibromofuran |
| SMILES | BrC1=CC=C(Br)O1 |
Chemical Identifiers
| 32460-00-7 | |
| 225.87 | |
| LACYYWKMIJOHLU-UHFFFAOYSA-N | |
| 2,5-dibromofuran |
| C4H2Br2O | |
| MFCD00191342 | |
| 10878846 | |
| BrC1=CC=C(Br)O1 |
Specifications
| 32460-00-7 | |
| Yellow | |
| C4H2Br2O | |
| 5 g | |
| LACYYWKMIJOHLU-UHFFFAOYSA-N | |
| 2,5-dibromofuran | |
| 10878846 | |
| ≥97.0% (GC) | |
| 2,5-Dibromofuran (stabilized with MgO) |
| 10°C | |
| 165°C | |
| MFCD00191342 | |
| 1993 | |
| BrC1=CC=C(Br)O1 | |
| 225.87 | |
| 225.87 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator