accessibility menu, dialog, popup

UserName

CAS RN 32460-00-7

OBrBr

CAS RN 32460-00-7

IUPAC Name: 2,5-dibromofuran
Synonyms: 2,5-dibromofuran
Molecular Weight (g/mol): 225.87
Molecular Formula: C4H2Br2O
InChi Key: LACYYWKMIJOHLU-UHFFFAOYSA-N
SMILES: BrC1=CC=C(Br)O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results