missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromofuran (stabilized with MgO) 97.0+%, TCI America™

Catalog No. D23665G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23665G 5 g
D236625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23665G Supplier TCI America Supplier No. D23665G
Only null left

Chemical Identifiers

CAS 32460-00-7
Molecular Formula C4H2Br2O
Molecular Weight (g/mol) 225.87
MDL Number MFCD00191342
InChI Key LACYYWKMIJOHLU-UHFFFAOYSA-N
PubChem CID 10878846
IUPAC Name 2,5-dibromofuran
SMILES BrC1=CC=C(Br)O1

Specifications

Melting Point 10°C
Color Yellow
Boiling Point 165°C
Quantity 5 g
UN Number 1993
Formula Weight 225.87
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dibromofuran (stabilized with MgO)
TSCA No
Recommended Storage Refrigerator