missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromobenzotrifluoride 98.0+%, TCI America™

Catalog No. D47695G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47695G 5 g
D476925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47695G Supplier TCI America Supplier No. D47695G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7657-09-2
Molecular Formula C7H3Br2F3
Molecular Weight (g/mol) 303.90
MDL Number MFCD00013554
InChI Key VWKFJAOCLPPQGR-UHFFFAOYSA-N
Synonym 1,4-dibromo-2-trifluoromethyl benzene, 2,5-dibromobenzotrifluoride, benzene, 1,4-dibromo-2-trifluoromethyl, 2,5-dibromo trifluoromethyl benzene, 1,4-dibromo-2-trifluoromethyl-benzene, toluene, 2,5-dibromo-alpha,alpha,alpha-trifluoro, pubchem9537, 2,5-dibromobenzotrifloride, 2,5-dibromo-, ,-trifluorotoluene
PubChem CID 82108
IUPAC Name 1,4-dibromo-2-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC(Br)=CC=C1Br

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 207°C
Quantity 5 g
Formula Weight 303.90
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,5-Dibromobenzotrifluoride