missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromobenzaldehyde, 97%

Catalog No. AAH3168906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3168906 5 g
AAH3168903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3168906 Supplier Thermo Scientific Chemicals Supplier No. H3168906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 74553-29-0 | C7H4Br2O | 263.916 g/mol

2,5-Dibromobenzaldehyde is used as intermediate, it is used to produce other chemicals, it can react with Ethane-1,2-diol to get 2-(2,5-Dibromo-phenyl)-[1,3]dioxolane. The reaction occurs with reagent p-toluenesulfonic acid, solvent toluene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dibromobenzaldehyde is used as intermediate, it is used to produce other chemicals, it can react with Ethane-1,2-diol to get 2-(2,5-Dibromo-phenyl)-[1,3]dioxolane. The reaction occurs with reagent p-toluenesulfonic acid, solvent toluene.

Solubility
Soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers

Chemical Identifiers

CAS 74553-29-0
Molecular Formula C7H4Br2O
Molecular Weight (g/mol) 263.916
MDL Number MFCD00462864
InChI Key BQBXKWGMPUCSQV-UHFFFAOYSA-N
Synonym benzaldehyde, 2,5-dibromo, 2,5-dibromo-benzaldehyde, zlchem 813, acmc-20a0sm, 2,5-di bromobenzaldehyde, ksc493q1h, 2,5-dibromobenzaldehyde
PubChem CID 13540069
IUPAC Name 2,5-dibromobenzaldehyde
SMILES C1=CC(=C(C=C1Br)C=O)Br

Specifications

Melting Point 88°C to 102°C
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 263.93
Percent Purity 97%
Chemical Name or Material 2,5-Dibromobenzaldehyde
Hazard Category H301-H314-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only