missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromoaniline, 97%

CAS: 3638-73-1 | C6H5Br2N | 250.921 g/mol

Supplier:  Thermo Scientific Chemicals A1911406

 View more versions of this product

Catalog No. AAA1911406


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,5-Dibromoaniline
51°C to 56°C
Characteristic
MFCD00007636
UN2811
2,5-dibromobenzenamine, benzenamine, 2,5-dibromo, 2,5-dibromophenylamine, rarechem fh 1w 0043, pubchem3452, 2,5-dibromo-phenylamine, aniline, 2,5-dibromo, 2,5-bis bromanyl aniline, acmc-209im6, 2,5-dibromoaniline
C1=CC(=C(C=C1Br)N)Br
250.921
250.93
3638-73-1
>110°C (230°F)
C6H5Br2N
5 g
1817548
WRTAZRGRFBCKBU-UHFFFAOYSA-N
2,5-dibromoaniline
77198
98%

Chemical Identifiers

3638-73-1
250.921
WRTAZRGRFBCKBU-UHFFFAOYSA-N
77198
C1=CC(=C(C=C1Br)N)Br
C6H5Br2N
MFCD00007636
2,5-dibromobenzenamine, benzenamine, 2,5-dibromo, 2,5-dibromophenylamine, rarechem fh 1w 0043, pubchem3452, 2,5-dibromo-phenylamine, aniline, 2,5-dibromo, 2,5-bis bromanyl aniline, acmc-209im6, 2,5-dibromoaniline
2,5-dibromoaniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P314-P330-P332+P313-P361-P363-P501c

H301+H311+H331-H315-H319-H335-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 222-865-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only