missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-p-xylene, 98+%

Catalog No. p-7043639
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0177614 25 g
AAL0177622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0177614 Supplier Thermo Scientific Chemicals Supplier No. L0177614

CAS: 1074-24-4 | C8H8Br2 | 263.96 g/mol

2,5-Dibromo-p-xylene is used as a liquid crystal material. It is also used as an intermediate in agrochemicals and in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dibromo-p-xylene is used as a liquid crystal material. It is also used as an intermediate in agrochemicals and in pharmaceuticals.

Solubility
Soluble in toluene, hot ethanol and hot methanol.Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and strong alkalis.

Chemical Identifiers

CAS 1074-24-4
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.96
MDL Number MFCD00000091
InChI Key QENIALCDPFDFHX-UHFFFAOYSA-N
Synonym 2,5-dibromo-p-xylene, benzene, 1,4-dibromo-2,5-dimethyl, p-xylene, 2,5-dibromo, 2,5-dibromo-1,4-dimethylbenzene, 2,5-dimethyl-1,4-dibromobenzene, 2,5-dibromo-4-xylene, 2,4-dibromobenzene, p-xylene,5-dibromo, pubchem14829, acmc-1c2tg
PubChem CID 66175
IUPAC Name 1,4-dibromo-2,5-dimethylbenzene
SMILES CC1=CC(Br)=C(C)C=C1Br

Specifications

Melting Point 72°C to 74°C
Quantity 25 g
Odor Odorless
Beilstein 2080433
Boiling Point 261°C
Solubility Information Soluble in toluene,hot ethanol and hot methanol.Insoluble in water.
Formula Weight 263.97
Percent Purity ≥98%
Chemical Name or Material 2,5-Dibromo-p-xylene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-038-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only