missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3-tetradecylthiophene 97.0+%, TCI America™

Catalog No. D52415G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52415G 5 g
D52411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52415G Supplier TCI America Supplier No. D52415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 160096-74-2
Molecular Formula C18H30Br2S
Molecular Weight (g/mol) 438.31
MDL Number MFCD30742898
InChI Key IYQPIBPAABQINF-UHFFFAOYSA-N
PubChem CID 101678000
IUPAC Name 2,5-dibromo-3-tetradecylthiophene
SMILES CCCCCCCCCCCCCCC1=C(Br)SC(Br)=C1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 438.31
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,5-Dibromo-3-tetradecylthiophene