accessibility menu, dialog, popup

UserName

CAS RN 160096-74-2

IUPAC Name: 2,5-dibromo-3-tetradecylthiophene
Synonyms: 2,5-dibromo-3-tetradecylthiophene
Molecular Weight (g/mol): 438.31
Molecular Formula: C18H30Br2S
InChi Key: IYQPIBPAABQINF-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCC1=C(Br)SC(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results