missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3-nitropyridine 98.0+%, TCI America™

Catalog No. D47021G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47021G 1 g
D47025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47021G Supplier TCI America Supplier No. D47021G
Only null left

Chemical Identifiers

CAS 15862-37-0
Molecular Formula C5H2Br2N2O2
Molecular Weight (g/mol) 281.891
MDL Number MFCD09266223
InChI Key OQKWPJCAKRVADO-UHFFFAOYSA-N
Synonym 2,5-dibromo-3-nitro-pyridine, 2,5-di bromo-3-nitro pyridine, pyridine, 2,5-dibromo-3-nitro, pubchem5505, acmc-1bp61, ksc495e2t, 2 pound not5-dibromo-3-nitropyridine
PubChem CID 298470
IUPAC Name 2,5-dibromo-3-nitropyridine
SMILES C1=C(C(=NC=C1Br)Br)[N+](=O)[O-]

Specifications

Melting Point 94°C
Color Yellow
Quantity 1 g
Formula Weight 281.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromo-3-nitropyridine