accessibility menu, dialog, popup

UserName

ONOBrNBr

CAS RN 15862-37-0

IUPAC Name: 2,5-dibromo-3-nitropyridine
Synonyms: 2,5-dibromo-3-nitropyridine
Molecular Weight (g/mol): 281.89
Molecular Formula: C5H2Br2N2O2
InChi Key: OQKWPJCAKRVADO-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Br)=CN=C1Br
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)