missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3-methylthiophene, 98%

Catalog No. p-7043779
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0269306 5 g
AAL0269314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0269306 Supplier Thermo Scientific Chemicals Supplier No. L0269306
Only null left

CAS: 13191-36-1 | C5H4Br2S | 255.955 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13191-36-1
Molecular Formula C5H4Br2S
Molecular Weight (g/mol) 255.955
MDL Number MFCD00015470
InChI Key IHFXZROPBCBLLG-UHFFFAOYSA-N
Synonym thiophene, 2,5-dibromo-3-methyl, 2,5-dibromo-3-methyl-thiophene, acmc-1bt3y, 2-p-nitrophenylthio ethanol, 2,5-dibrom-3-methylthiophen, 3-methyl-2,5-dibromothiophene, 2,5-dibromo-3-methylthiophene
PubChem CID 83219
IUPAC Name 2,5-dibromo-3-methylthiophene
SMILES CC1=C(SC(=C1)Br)Br

Specifications

Density 1.972
Boiling Point 226°C to 230°C
Refractive Index 1.613
Quantity 5 g
Beilstein 108894
Sensitivity Light sensitive
Formula Weight 255.97
Percent Purity 98%
Chemical Name or Material 2,5-Dibromo-3-methylthiophene
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 236-147-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only