accessibility menu, dialog, popup

UserName

CAS RN 13191-36-1

IUPAC Name: 2,5-dibromo-3-methylthiophene
Synonyms: 2,5-dibromo-3-methylthiophene
Molecular Weight (g/mol): 255.96
Molecular Formula: C5H4Br2S
InChi Key: IHFXZROPBCBLLG-UHFFFAOYSA-N
SMILES: CC1=C(Br)SC(Br)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)