missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3-hexadecylthiophene 96.0+%, TCI America™

Catalog No. p-7128703
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
D52421G 1 g
D52425G 5 g
2 options

Catalog No. D52421G

Supplier: TCI America D52421G

Only null left

Chemical Identifiers

CAS 178452-13-6
Molecular Formula C20H34Br2S
Molecular Weight (g/mol) 466.36
MDL Number MFCD30742899
InChI Key KBLOMHRWQCLPTC-UHFFFAOYSA-N
PubChem CID 101022660
IUPAC Name 2,5-dibromo-3-hexadecylthiophene
SMILES CCCCCCCCCCCCCCCCC1=C(Br)SC(Br)=C1

Specifications

Melting Point 36°C
Color White-Yellow
Quantity 1 g
Formula Weight 466.36
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromo-3-hexadecylthiophene
TSCA No
Recommended Storage Refrigerator