accessibility menu, dialog, popup

UserName

H3CSBrBr

CAS RN 178452-13-6

IUPAC Name: 2,5-dibromo-3-hexadecylthiophene
Synonyms: 2,5-dibromo-3-hexadecylthiophene
Molecular Weight (g/mol): 466.36
Molecular Formula: C20H34Br2S
InChi Key: KBLOMHRWQCLPTC-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCC1=C(Br)SC(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results