missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dibromo-3-hexadecylthiophene 96.0+%, TCI America™
Specifications
Specifications
| Melting Point | 36°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 466.36 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,5-Dibromo-3-hexadecylthiophene |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |