missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-1,1-dimethyl-3,4-diphenylsilole 98.0+%, TCI America™

Catalog No. D46311G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46311G 1 g
D46315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46311G Supplier TCI America Supplier No. D46311G
Only null left

Chemical Identifiers

CAS 686290-22-2
Molecular Formula C18H16Br2Si
Molecular Weight (g/mol) 420.22
MDL Number MFCD28384141
InChI Key XLVXIPAEFXBVBF-UHFFFAOYSA-N
Synonym 2,5-Dibromo-1,1-dimethyl-3,4-diphenylsilacyclopenta-2,4-diene
PubChem CID 11729466
IUPAC Name 2,5-dibromo-1,1-dimethyl-3,4-diphenyl-1H-silole
SMILES C[Si]1(C)C(Br)=C(C(=C1Br)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 163°C
Color Yellow
Quantity 1 g
Formula Weight 420.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromo-1,1-dimethyl-3,4-diphenylsilole
TSCA No
Recommended Storage Refrigerator