accessibility menu, dialog, popup

UserName

H3CSiBrBrCH3

CAS RN 686290-22-2

IUPAC Name: 2,5-dibromo-1,1-dimethyl-3,4-diphenyl-1H-silole
Synonyms: 2,5-dibromo-1,1-dimethyl-3,4-diphenylsilole
Molecular Weight (g/mol): 420.22
Molecular Formula: C18H16Br2Si
InChi Key: XLVXIPAEFXBVBF-UHFFFAOYSA-N
SMILES: C[Si]1(C)C(Br)=C(C(=C1Br)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)