missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dibenzylidenecyclopentanone 98.0+%, TCI America™
$165.90 - $831.37
Chemical Identifiers
| CAS | 895-80-7 |
|---|---|
| Molecular Formula | C19H16O |
| Molecular Weight (g/mol) | 260.336 |
| MDL Number | MFCD00019321 |
| InChI Key | CVTOCKKPVXJIJK-SIJTWYJSSA-N |
| PubChem CID | 6436781 |
| IUPAC Name | (2Z,5E)-2,5-dibenzylidenecyclopentan-1-one |
| SMILES | C1CC(=CC2=CC=CC=C2)C(=O)C1=CC3=CC=CC=C3 |
Chemical Identifiers
| 895-80-7 | |
| 260.336 | |
| CVTOCKKPVXJIJK-SIJTWYJSSA-N | |
| (2Z,5E)-2,5-dibenzylidenecyclopentan-1-one |
| C19H16O | |
| MFCD00019321 | |
| 6436781 | |
| C1CC(=CC2=CC=CC=C2)C(=O)C1=CC3=CC=CC=C3 |
Specifications
| 895-80-7 | |
| Yellow | |
| MFCD00019321 | |
| CVTOCKKPVXJIJK-SIJTWYJSSA-N | |
| (2Z,5E)-2,5-dibenzylidenecyclopentan-1-one | |
| 6436781 | |
| ≥98.0% (HPLC) | |
| 2,5-Dibenzylidenecyclopentanone |
| 192°C | |
| C19H16O | |
| 5 g | |
| C1CC(=CC2=CC=CC=C2)C(=O)C1=CC3=CC=CC=C3 | |
| 260.336 | |
| 260.34 | |
| Crystalline Powder |
Safety and Handling
TSCA : No