accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 895-80-7

O(E)(E)

CAS RN 895-80-7

IUPAC Name: (2E,5E)-2,5-bis(phenylmethylidene)cyclopentan-1-one
Synonyms: (2E,5E)-2,5-bis(phenylmethylidene)cyclopentan-1-one
Molecular Weight (g/mol): 260.34
Molecular Formula: C19H16O
InChi Key: CVTOCKKPVXJIJK-HBKJEHTGSA-N
SMILES: O=C1\C(CC\C1=C/C1=CC=CC=C1)=C\C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.