missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diaminopyridine, 97%

Catalog No. p-4812688
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC360690010 1 g
AC360690050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC360690010 Supplier Thermo Scientific Chemicals Supplier No. 360690010
Only null left

CAS: 4318-76-7 | C5H7N3 | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4318-76-7
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.13
InChI Key MIROPXUFDXCYLG-UHFFFAOYSA-N
Synonym 2,5-diaminopyridine, 2,5-pyridinediamine, pyridine, 2,5-diamino, diaminopyridine van, unii-h6olz060hi, h6olz060hi, da1, pyridine,5-diamino, 2,5-diamino pyridine, 2,5-diamino-pyridine
PubChem CID 20314
IUPAC Name pyridine-2,5-diamine
SMILES C1=CC(=NC=C1N)N

Specifications

Boiling Point 180.0°C (12.0 mmHg)
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 109.13
Percent Purity 97%
Chemical Name or Material 2, 5-Diaminopyridine
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only