accessibility menu, dialog, popup

UserName

CAS RN 4318-76-7

NH2NH2N

CAS RN 4318-76-7

IUPAC Name: pyridine-2,5-diamine
Synonyms: 2,5-diaminopyridine
Molecular Weight (g/mol): 109.13
Molecular Formula: C5H7N3
InChi Key: MIROPXUFDXCYLG-UHFFFAOYSA-N
SMILES: NC1=CC=C(N)N=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)