missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diaminohydroquinone dihydrochloride, 97%

Catalog No. AAB2196901
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2196901 0.25 g
AAB2196903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2196901 Supplier Thermo Scientific Chemicals Supplier No. B2196901
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 24171-03-7 | C6H10Cl2N2O2 | 213.058 g/mol

2,5-Diaminohydroquinone dihydrochloride is used in the preparation of polymers, synthetically cross-linked polyimide and silica hybrid films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Diaminohydroquinone dihydrochloride is used in the preparation of polymers, synthetically cross-linked polyimide and silica hybrid films.

Notes
Air sensitive. It may discolor to black-violet on storage. Store away from strong oxidizing agents, air.

Chemical Identifiers

CAS 24171-03-7
Molecular Formula C6H10Cl2N2O2
Molecular Weight (g/mol) 213.058
MDL Number MFCD00239416
InChI Key NILKAWPWTYPHAH-UHFFFAOYSA-N
Synonym 2,5-diaminobenzene-1,4-diol dihydrochloride, 2,5-diamino-1,4-dihydroxybenzene dihydrochloride, 2,5-diaminohydroquinone dihydrochloride, acmc-20aobi, 1,4-benzenediol, 2,5-diamino-, dihydrochloride, 2,5-diaminobenzene-1,4-diol, chloride, chloride, 1,4-benzenediol,2,5-diamino-,hydrochloride 1:2, 2,5-diaminobenzene-1,4-diol-hydrogen chloride 1/2, 2,5-diaminohydroquinone dihydrochloride, technical at
PubChem CID 16212088
IUPAC Name 2,5-diaminobenzene-1,4-diol;dihydrochloride
SMILES C1=C(C(=CC(=C1O)N)O)N.Cl.Cl

Specifications

Melting Point >300°C
Odor Odorless
Quantity 0.25 g
Sensitivity Air Sensitive
Formula Weight 213.06
Percent Purity 97%
Chemical Name or Material 2,5-Diaminohydroquinone dihydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only