missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diamino-4,6-dihydroxypyrimidine Hydrochloride 98.0+%, TCI America™

Catalog No. D26345G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26345G 5 g
D263425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26345G Supplier TCI America Supplier No. D26345G

Chemical Identifiers

CAS 56830-58-1
Molecular Formula C4H7ClN4O2
Molecular Weight (g/mol) 178.576
MDL Number MFCD00082893
InChI Key IIVYOQGIZSPVFZ-UHFFFAOYSA-N
Synonym 2,5-diamino-4,6-dihydroxypyrimidine hydrochloride, 2,5-diaminopyrimidine-4,6-diol hydrochloride, 2,5-diamino-4,6-dihydroxypyrimidinehydrochloride, 2,5-diamino-6-hydroxy-3h-pyrimidin-4-one hydrochloride, diaminopyrimidine-4,6-diol hydrochloride, 2,5-diamino-4,6-dihydroxypyrimidine hcl, 2,5-diamino-4-hydroxy-1h-pyrimidin-6-one hydrochloride, 4 1h-pyrimidinone, 2,5-diamino-6-hydroxy-, monohydrochloride, 2,5-diaminopyrimidine-4,6-diol, chloride, 2,5-diamino-6-hydroxy-3h-pyrimidin-4-one; hydrochloride
PubChem CID 2733980
IUPAC Name 2,5-diamino-4-hydroxy-1H-pyrimidin-6-one;hydrochloride
SMILES C1(=C(N=C(NC1=O)N)O)N.Cl

Specifications

Color Red-White
Quantity 5 g
Formula Weight 178.58
Percent Purity ≥98.0% (T,HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Diamino-4,6-dihydroxypyrimidine Hydrochloride