missing translation for 'onlineSavingsMsg'
Learn More

2,5-Di-tert-butylhydroquinone 98.0+%, TCI America™

Catalog No. D094025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D094025G 25 g
D0940500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D094025G Supplier TCI America Supplier No. D094025G

Chemical Identifiers

CAS 88-58-4
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.33
MDL Number MFCD00008825
InChI Key JZODKRWQWUWGCD-UHFFFAOYSA-N
Synonym 2,5-di-tert-butylhydroquinone, 2,5-di-tert-butylbenzene-1,4-diol, dibug, dtbhq, dybug, santovar o, di-t-butylhydroquinone, 2,5-di-tert-butylquinol, nonflex alba, 2,5-di-t-butylhydroquinone
PubChem CID 2374
ChEBI CHEBI:41094
IUPAC Name 2,5-di-tert-butylbenzene-1,4-diol
SMILES CC(C)(C)C1=CC(O)=C(C=C1O)C(C)(C)C

Specifications

Melting Point 215°C
Boiling Point 321°C
Quantity 25 g
Formula Weight 222.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Di-tert-butylhydroquinone
EINECSNumber (3)-0553
RTECSNumber MX5160000
TSCA Yes