missing translation for 'onlineSavingsMsg'
Learn More

2,5-Di-tert-butylhydroquinone, 99%

Catalog No. p-3778165
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC150840010 1 kg Plastic bottle
AC150840025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC150840010 Supplier Thermo Scientific Chemicals Supplier No. 150840010
Only null left

CAS: 88-58-4 | C14H22O2 | 222.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88-58-4
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.33
MDL Number MFCD00008825
InChI Key JZODKRWQWUWGCD-UHFFFAOYSA-N
Synonym 2,5-di-tert-butylhydroquinone, 2,5-di-tert-butylbenzene-1,4-diol, dibug, dtbhq, dybug, santovar o, di-t-butylhydroquinone, 2,5-di-tert-butylquinol, nonflex alba, 2,5-di-t-butylhydroquinone
PubChem CID 2374
ChEBI CHEBI:41094
IUPAC Name 2,5-ditert-butylbenzene-1,4-diol
SMILES CC(C)(C)C1=CC(O)=C(C=C1O)C(C)(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 212°C to 218°C
Color Beige to Beige-Pink
Density 216g/mL
Flash Point 216°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Quantity 1 kg
Solubility Information Solubility in water: insoluble in water. Other solubilities: insoluble in aqueous alkali,soluble in acetone,alcohol and benzene,practically insoluble in toluene and hexane
Formula Weight 222.33
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 5-Di-tert-butylhydroquinone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-841-8

RUO – Research Use Only