missing translation for 'onlineSavingsMsg'
Learn More

2,5-Di-tert-butyl-4-methoxyphenol, 97%

Catalog No. AAH3505306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3505306 5 g
AAH3505314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3505306 Supplier Thermo Scientific Chemicals Supplier No. H3505306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1991-52-2 | C15H24O2 | 236.355 g/mol

2,5-Di-tert-butyl-4-methoxyphenol is used as a non-tumorigenic compound in contrast to DBHQ.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Di-tert-butyl-4-methoxyphenol is used as a non-tumorigenic compound in contrast to DBHQ.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1991-52-2
Molecular Formula C15H24O2
Molecular Weight (g/mol) 236.355
MDL Number MFCD00274238
InChI Key FLLRQABPKFCXSO-UHFFFAOYSA-N
Synonym 2,5-di-tert-butyl-4-methoxyphenol, 2,5-di-tert-butyl-4-hydroxyanisole, ccris 5220, unii-593e9t2mwh, phenol,2,5-bis 1,1-dimethylethyl-4-methoxy, acmc-1bqnv, spectrum1505039, 2,5-di-t-butyl-4-methoxyphenol, 2,5-di-t-butyl-4-hydroxyanisole, 2,5-di-tert-butyl4-hydroxyanisole
PubChem CID 74812
IUPAC Name 2,5-ditert-butyl-4-methoxyphenol
SMILES CC(C)(C)C1=CC(=C(C=C1OC)C(C)(C)C)O

Specifications

Melting Point 99°C to 102°C
Color White to Cream
Quantity 5 g
Formula Weight 236.36
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Di-tert-butyl-4-methoxyphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-873-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only