missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Di-tert-amylhydroquinone 93.0+%, TCI America™
$69.31 - $500.54
Chemical Identifiers
| CAS | 79-74-3 |
|---|---|
| Molecular Formula | C16H26O2 |
| Molecular Weight (g/mol) | 250.38 |
| MDL Number | MFCD00027029 |
| InChI Key | CZNRFEXEPBITDS-UHFFFAOYSA-N |
| Synonym | 2,5-Di-tert-pentylhydroquinone |
| PubChem CID | 6610 |
| IUPAC Name | 2,5-bis(2-methylbutan-2-yl)benzene-1,4-diol |
| SMILES | CCC(C)(C)C1=CC(O)=C(C=C1O)C(C)(C)CC |
Chemical Identifiers
| 79-74-3 | |
| 250.38 | |
| CZNRFEXEPBITDS-UHFFFAOYSA-N | |
| 6610 | |
| CCC(C)(C)C1=CC(O)=C(C=C1O)C(C)(C)CC |
| C16H26O2 | |
| MFCD00027029 | |
| 2,5-Di-tert-pentylhydroquinone | |
| 2,5-bis(2-methylbutan-2-yl)benzene-1,4-diol |
Specifications
| 79-74-3 | |
| C16H26O2 | |
| 25 g | |
| CZNRFEXEPBITDS-UHFFFAOYSA-N | |
| 2,5-bis(2-methylbutan-2-yl)benzene-1,4-diol | |
| 6610 | |
| ≥93.0% (GC) | |
| 2,5-Di-tert-amylhydroquinone |
| 179°C | |
| MFCD00027029 | |
| 2,5-Di-tert-pentylhydroquinone | |
| CCC(C)(C)C1=CC(O)=C(C=C1O)C(C)(C)CC | |
| 250.38 | |
| 250.38 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-0553
RTECSNumber : MX6300000
TSCA : Yes