missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(5-Chloro-2-benzotriazolyl)-6-tert-butyl-p-cresol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 140°C |
| Color | Yellow |
| Quantity | 500 g |
| Formula Weight | 315.80 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-(5-Chloro-2-benzotriazolyl)-6-tert-butyl-p-cresol |
Safety and Handling
| EINECSNumber | (5)-0545 |
| TSCA | Yes |