missing translation for 'onlineSavingsMsg'
Learn More

2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol 98.0+%, TCI America™

Catalog No. p-7118839
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1081100MG 100 mg
B10811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1081100MG Supplier TCI America Supplier No. B1081100MG
Only null left

Chemical Identifiers

CAS 14337-53-2
Molecular Formula C15H17BrN4O
Molecular Weight (g/mol) 349.23
MDL Number MFCD00006255
InChI Key LYVSIKOGJUDRBK-QGOAFFKASA-N
Synonym 2-5-bromo-2-pyridylazo-5-diethylamino phenol, 2-5-bromopyridin-2-yl diazenyl-5-diethylamino phenol, bromo-padap, 2-5-bromo-2-pyridylazo-5-diethylaminophenol, phenol, 2-5-bromo-2-pyridinyl azo-5-diethylamino, 2-4-diethylamino-2-hydroxyphenylazo-5-bromopyridine, hnvcxdavehoibp-vhebqxmusa-n, 2-5-bromo-2-pyridylazo-5-diethylamino-phenol, 5-diethylamino-2-5-bromo 2-pyridyl diazenyl phenol, e-2-5-bromopyridin-2-yl diazenyl-5-diethylamino phenol
PubChem CID 5911417
IUPAC Name (6E)-6-[2-(5-bromopyridin-2-yl)hydrazin-1-ylidene]-3-(diethylamino)cyclohexa-2,4-dien-1-one
SMILES CCN(CC)C1=CC(=O)\C(C=C1)=N\NC1=CC=C(Br)C=N1

Specifications

Melting Point 157°C
Quantity 100 mg
Formula Weight 349.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol