accessibility menu, dialog, popup

UserName

CAS RN 14337-53-2

IUPAC Name: (6E)-6-[2-(5-bromopyridin-2-yl)hydrazin-1-ylidene]-3-(diethylamino)cyclohexa-2,4-dien-1-one
Synonyms: (6E)-6-[2-(5-bromopyridin-2-yl)hydrazin-1-ylidene]-3-(diethylamino)cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 349.23
Molecular Formula: C15H17BrN4O
InChi Key: LYVSIKOGJUDRBK-QGOAFFKASA-N
SMILES: CCN(CC)C1=CC(=O)\C(C=C1)=N\NC1=CC=C(Br)C=N1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)