missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol, 98%

CAS: 14337-53-2 | C15H17BrN4O | 349.23 g/mol

$303.11 - $303.11

Chemical Identifiers

CAS 14337-53-2
Molecular Formula C15H17BrN4O
Molecular Weight (g/mol) 349.23
MDL Number MFCD00006255
InChI Key LYVSIKOGJUDRBK-QGOAFFKASA-N
Synonym 2-5-bromo-2-pyridylazo-5-diethylamino phenol, 2-5-bromopyridin-2-yl diazenyl-5-diethylamino phenol, bromo-padap, 2-5-bromo-2-pyridylazo-5-diethylaminophenol, phenol, 2-5-bromo-2-pyridinyl azo-5-diethylamino, 2-4-diethylamino-2-hydroxyphenylazo-5-bromopyridine, hnvcxdavehoibp-vhebqxmusa-n, 2-5-bromo-2-pyridylazo-5-diethylamino-phenol, 5-diethylamino-2-5-bromo 2-pyridyl diazenyl phenol, e-2-5-bromopyridin-2-yl diazenyl-5-diethylamino phenol
PubChem CID 5911417
IUPAC Name (6E)-6-[(5-bromopyridin-2-yl)hydrazinylidene]-3-(diethylamino)cyclohexa-2,4-dien-1-one
SMILES CCN(CC)C1=CC(=O)\C(C=C1)=N\NC1=CC=C(Br)C=N1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC403220010
SDS View Documents
Thermo Scientific Chemicals
403220010
1 g Glass bottle
Each for $303.11
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14337-53-2
349.23
LYVSIKOGJUDRBK-QGOAFFKASA-N
5911417
CCN(CC)C1=CC(=O)\C(C=C1)=N\NC1=CC=C(Br)C=N1
C15H17BrN4O
MFCD00006255
2-5-bromo-2-pyridylazo-5-diethylamino phenol, 2-5-bromopyridin-2-yl diazenyl-5-diethylamino phenol, bromo-padap, 2-5-bromo-2-pyridylazo-5-diethylaminophenol, phenol, 2-5-bromo-2-pyridinyl azo-5-diethylamino, 2-4-diethylamino-2-hydroxyphenylazo-5-bromopyridine, hnvcxdavehoibp-vhebqxmusa-n, 2-5-bromo-2-pyridylazo-5-diethylamino-phenol, 5-diethylamino-2-5-bromo 2-pyridyl diazenyl phenol, e-2-5-bromopyridin-2-yl diazenyl-5-diethylamino phenol
(6E)-6-[(5-bromopyridin-2-yl)hydrazinylidene]-3-(diethylamino)cyclohexa-2,4-dien-1-one

Specifications

14337-53-2
100.0
Red
97.5% min. (HPLC)
C15H17BrN4O
1 g
LYVSIKOGJUDRBK-QGOAFFKASA-N
(6E)-6-[(5-bromopyridin-2-yl)hydrazinylidene]-3-(diethylamino)cyclohexa-2,4-dien-1-one
5911417
98%
2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol
99.0
155.0°C to 160.0°C
Authentic
Glass bottle
MFCD00006255
2-5-bromo-2-pyridylazo-5-diethylamino phenol, 2-5-bromopyridin-2-yl diazenyl-5-diethylamino phenol, bromo-padap, 2-5-bromo-2-pyridylazo-5-diethylaminophenol, phenol, 2-5-bromo-2-pyridinyl azo-5-diethylamino, 2-4-diethylamino-2-hydroxyphenylazo-5-bromopyridine, hnvcxdavehoibp-vhebqxmusa-n, 2-5-bromo-2-pyridylazo-5-diethylamino-phenol, 5-diethylamino-2-5-bromo 2-pyridyl diazenyl phenol, e-2-5-bromopyridin-2-yl diazenyl-5-diethylamino phenol
CCN(CC)C1=CC(=O)\C(C=C1)=N\NC1=CC=C(Br)C=N1
349.23
349.22
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 238-286-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only