missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(trimethylstannyl)thieno[3,2-b]thiophene 98.0+%, TCI America™

Catalog No. p-7123602
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4536200MG 200 mg
B45361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4536200MG Supplier TCI America Supplier No. B4536200MG
Only null left

Chemical Identifiers

CAS 469912-82-1
Molecular Formula C12H20S2Sn2
Molecular Weight (g/mol) 465.83
MDL Number MFCD22200503
InChI Key HDZULVYGCRXVNQ-UHFFFAOYSA-N
PubChem CID 12978043
IUPAC Name trimethyl[5-(trimethylstannyl)thieno[3,2-b]thiophen-2-yl]stannane
SMILES C[Sn](C)(C)C1=CC2=C(S1)C=C(S2)[Sn](C)(C)C

Specifications

Melting Point 129°C
Quantity 200 mg
UN Number 3146
Formula Weight 465.83
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Bis(trimethylstannyl)thieno[3,2-b]thiophene