accessibility menu, dialog, popup

UserName

CAS RN 469912-82-1

H3CSnCH3CH3SSSnCH3CH3CH3

CAS RN 469912-82-1

IUPAC Name: trimethyl[5-(trimethylstannyl)thieno[3,2-b]thiophen-2-yl]stannane
Synonyms: trimethyl[5-(trimethylstannyl)thieno[3,2-b]thiophen-2-yl]stannane
Molecular Weight (g/mol): 465.83
Molecular Formula: C12H20S2Sn2
InChi Key: HDZULVYGCRXVNQ-UHFFFAOYSA-N
SMILES: C[Sn](C)(C)C1=CC2=C(S1)C=C(S2)[Sn](C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results