missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(trifluoromethyl)benzoic acid, 98%

Catalog No. AAH2618406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2618406 5 g
AAH2618403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2618406 Supplier Thermo Scientific Chemicals Supplier No. H2618406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 42580-42-7 | C9H4F6O2 | 258.119 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42580-42-7
Molecular Formula C9H4F6O2
Molecular Weight (g/mol) 258.119
MDL Number MFCD00013249
InChI Key PINBPLCVZSKLTF-UHFFFAOYSA-N
PubChem CID 736187
ChEBI CHEBI:60697
IUPAC Name 2,5-bis(trifluoromethyl)benzoic acid
SMILES C1=CC(=C(C=C1C(F)(F)F)C(=O)O)C(F)(F)F

Specifications

Melting Point 77°C to 80°C
Quantity 5 g
Synonym 2,5-bis trifluoromethyl benzoic acid, 2,5-di trifluoromethyl benzoic acid, 2,5-bis trifluoroformyl benzoic acid, pubchem8492, acmc-1amwk, rarechem al bo 1002, 2,5-bis-trifluoromethyl-benzoic acid, 2,5-bis trifluoromethyl benzoic acid., benzoic acid,2,5-bis trifluoromethyl, benzoic acid, 2,5-bis trifluoromethyl
Formula Weight 258.12
Percent Purity 98%
Chemical Name or Material 2,5-Bis(trifluoromethyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only