missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(trifluoromethyl)aniline, 99%

CAS: 328-93-8 | C8H5F6N | 229.13 g/mol

$220.66 - $854.41

Chemical Identifiers

CAS 328-93-8
Molecular Formula C8H5F6N
Molecular Weight (g/mol) 229.13
MDL Number MFCD00074940
InChI Key XWMVIJUAZAEWIE-UHFFFAOYSA-N
Synonym 2,5-bis trifluoromethyl aniline, 2,5-di trifluoromethyl aniline, 2,5-bis-trifluoromethyl aniline, 2,5-bis-trifluoromethylaniline, benzenamine, 2,5-bis trifluoromethyl, 2,5-bis trifluoromethyl-aniline, 2,5-bis trifluoromethyl benzenamine, 1-amino-2,5-bis trifluoromethyl benzene, 2,5-bis trifluoromethyl phenylamine, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-2,5-xylidine
PubChem CID 719818
IUPAC Name 2,5-bis(trifluoromethyl)aniline
SMILES NC1=CC(=CC=C1C(F)(F)F)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1470606
View Documents
Thermo Scientific Chemicals
L1470606
5 g
Each for $220.66
Only null left
 
AAL1470614
View Documents
Thermo Scientific Chemicals
L1470614
25 g
Each for $854.41
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

328-93-8
229.13
XWMVIJUAZAEWIE-UHFFFAOYSA-N
719818
NC1=CC(=CC=C1C(F)(F)F)C(F)(F)F
C8H5F6N
MFCD00074940
2,5-bis trifluoromethyl aniline, 2,5-di trifluoromethyl aniline, 2,5-bis-trifluoromethyl aniline, 2,5-bis-trifluoromethylaniline, benzenamine, 2,5-bis trifluoromethyl, 2,5-bis trifluoromethyl-aniline, 2,5-bis trifluoromethyl benzenamine, 1-amino-2,5-bis trifluoromethyl benzene, 2,5-bis trifluoromethyl phenylamine, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-2,5-xylidine
2,5-bis(trifluoromethyl)aniline

Specifications

328-93-8
70°C to 71°C (15 mmHg)
Recognizable
1.4338
5 g
2653046
XWMVIJUAZAEWIE-UHFFFAOYSA-N
2,5-bis(trifluoromethyl)aniline
719818
99%
1.467
71°C (159°F)
C8H5F6N
MFCD00074940
UN2810
2,5-bis trifluoromethyl aniline, 2,5-di trifluoromethyl aniline, 2,5-bis-trifluoromethyl aniline, 2,5-bis-trifluoromethylaniline, benzenamine, 2,5-bis trifluoromethyl, 2,5-bis trifluoromethyl-aniline, 2,5-bis trifluoromethyl benzenamine, 1-amino-2,5-bis trifluoromethyl benzene, 2,5-bis trifluoromethyl phenylamine, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-2,5-xylidine
NC1=CC(=CC=C1C(F)(F)F)C(F)(F)F
229.13
229.13
2,5-Bis(trifluoromethyl)aniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H373-H227-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P210-P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H312+H332-H315-H319-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only