missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(4-biphenylyl)thiophene, TCI America™

Catalog No. B34411G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B34411G Supplier TCI America Supplier No. B34411G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56316-86-0
Molecular Formula C28H20S
Molecular Weight (g/mol) 388.528
InChI Key WBSMVAWETYTHTA-UHFFFAOYSA-N
Synonym BP1T
PubChem CID 4286041
IUPAC Name 2,5-bis(4-phenylphenyl)thiophene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(S3)C4=CC=C(C=C4)C5=CC=CC=C5

Specifications

Melting Point 337°C
Color Yellow
Quantity 1 g
Formula Weight 388.53
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Bis(4-biphenylyl)thiophene