accessibility menu, dialog, popup

UserName

CAS RN 56316-86-0

IUPAC Name: 2,5-bis({[1,1'-biphenyl]-4-yl})thiophene
Synonyms: 2,5-bis({[1,1'-biphenyl]-4-yl})thiophene
Molecular Weight (g/mol): 388.53
Molecular Formula: C28H20S
InChi Key: WBSMVAWETYTHTA-UHFFFAOYSA-N
SMILES: S1C(=CC=C1C1=CC=C(C=C1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results