missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(3,7-dimethyloctyloxy)terephthalaldehyde, 98%

Catalog No. AAH51067MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51067MD 250 mg
AAH5106703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH51067MD Supplier Thermo Scientific Chemicals Supplier No. H51067MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 325461-35-6 | C28H46O4 | 446.672 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 325461-35-6
Molecular Formula C28H46O4
Molecular Weight (g/mol) 446.672
MDL Number MFCD03427237
InChI Key JMACLBZCZVVLDQ-UHFFFAOYSA-N
Synonym 2,5-bis 3,7-dimethyloctyloxy terephthalaldehyde, 2,5-bis 3,7-dimethyloctoxy terephthalaldehyde, 2,5-bis-3,7-dimethyloctyloxy terephthalaldehyde, 2,5-bis 3,7-dimethyloctyl oxy benzene-1,4-dicarbaldehyde
PubChem CID 3653721
IUPAC Name 2,5-bis(3,7-dimethyloctoxy)terephthalaldehyde
SMILES CC(C)CCCC(C)CCOC1=CC(=C(C=C1C=O)OCCC(C)CCCC(C)C)C=O

Specifications

Quantity 250 mg
Formula Weight 446.66
Percent Purity 98%
Chemical Name or Material 2,5-Bis(3,7-dimethyloctyloxy)terephthalaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only