missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Bis(2-ethylhexyl)-3,6-di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione 98.0+%, TCI America™
Supplier: TCI America B54321G
Specifications
| 2,5-Bis(2-ethylhexyl)-3,6-di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione | |
| C30H40N2O2S2 | |
| 1 g | |
| CCCCC(CC)CN1C(=O)C2=C(N(CC(CC)CCCC)C(=O)C2=C1C1=CC=CS1)C1=CC=CS1 | |
| 524.78 | |
| 524.78 | |
| Crystalline Powder |
| 1185885-86-2 | |
| MFCD22200076 | |
| BTJNHAWTSFHBBN-UHFFFAOYNA-N | |
| 2,5-bis(2-ethylhexyl)-3,6-bis(thiophen-2-yl)-1H,2H,4H,5H-pyrrolo[3,4-c]pyrrole-1,4-dione | |
| 58517362 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 1185885-86-2 | |
| 524.78 | |
| BTJNHAWTSFHBBN-UHFFFAOYNA-N | |
| 2,5-bis(2-ethylhexyl)-3,6-bis(thiophen-2-yl)-1H,2H,4H,5H-pyrrolo[3,4-c]pyrrole-1,4-dione |
| C30H40N2O2S2 | |
| MFCD22200076 | |
| 58517362 | |
| CCCCC(CC)CN1C(=O)C2=C(N(CC(CC)CCCC)C(=O)C2=C1C1=CC=CS1)C1=CC=CS1 |
Safety and Handling
TSCA : No