missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(2-ethylhexyl)-3,6-di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione 98.0+%, TCI America™

Catalog No. p-7124074
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5432200MG 200 mg
B54321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5432200MG Supplier TCI America Supplier No. B5432200MG
Only null left

Chemical Identifiers

CAS 1185885-86-2
Molecular Formula C30H40N2O2S2
Molecular Weight (g/mol) 524.78
MDL Number MFCD22200076
InChI Key BTJNHAWTSFHBBN-UHFFFAOYNA-N
PubChem CID 58517362
IUPAC Name 2,5-bis(2-ethylhexyl)-3,6-bis(thiophen-2-yl)-1H,2H,4H,5H-pyrrolo[3,4-c]pyrrole-1,4-dione
SMILES CCCCC(CC)CN1C(=O)C2=C(N(CC(CC)CCCC)C(=O)C2=C1C1=CC=CS1)C1=CC=CS1

Specifications

Quantity 200 mg
Formula Weight 524.78
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Bis(2-ethylhexyl)-3,6-di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione