missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Trifluoromethylphenyl)ethylamine 98.0+%, TCI America™

Catalog No. T32785G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32785G 5 g
T32781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32785G Supplier TCI America Supplier No. T32785G

Chemical Identifiers

CAS 775-00-8
Molecular Formula C9H10F3N
Molecular Weight (g/mol) 189.181
MDL Number MFCD01529875
InChI Key HMFOBPNVAAAACP-UHFFFAOYSA-N
Synonym 4-trifluoromethylphenethylamine, 2-4-trifluoromethyl-phenyl-ethylamine, 2-4-trifluoromethyl phenyl ethanamine, 4-trifluoromethyl phenethylamine, 2-4-trifluoromethyl-phenyl ethylamine, 2-4-trifluoromethylphenyl ethylamine, 2-4-trifluoromethyl phenyl ethan-1-amine, 2-4-trifluoromethyl phenyl ethylamine, 2-4-trifluoromethyl phenyl ethyl amine
PubChem CID 1988106
IUPAC Name 2-[4-(trifluoromethyl)phenyl]ethanamine
SMILES C1=CC(=CC=C1CCN)C(F)(F)F

Specifications

Color Colorless
Boiling Point 90°C
Quantity 5 g
UN Number 2735
Formula Weight 189.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(4-Trifluoromethylphenyl)ethylamine
TSCA No
Recommended Storage Refrigerator