missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[4-(Trifluoromethyl)phenyl]pyridine 98.0+%, TCI America™
$211.77 - $635.31
Chemical Identifiers
| CAS | 203065-88-7 |
|---|---|
| Molecular Formula | C12H8F3N |
| Molecular Weight (g/mol) | 223.20 |
| MDL Number | MFCD08235168 |
| InChI Key | KGZSSIFFYUBVOX-UHFFFAOYSA-N |
| PubChem CID | 10933075 |
| IUPAC Name | 2-[4-(trifluoromethyl)phenyl]pyridine |
| SMILES | FC(F)(F)C1=CC=C(C=C1)C1=CC=CC=N1 |
Chemical Identifiers
| 203065-88-7 | |
| 223.20 | |
| KGZSSIFFYUBVOX-UHFFFAOYSA-N | |
| 2-[4-(trifluoromethyl)phenyl]pyridine |
| C12H8F3N | |
| MFCD08235168 | |
| 10933075 | |
| FC(F)(F)C1=CC=C(C=C1)C1=CC=CC=N1 |
Specifications
| 203065-88-7 | |
| White-Yellow | |
| MFCD08235168 | |
| KGZSSIFFYUBVOX-UHFFFAOYSA-N | |
| 2-[4-(trifluoromethyl)phenyl]pyridine | |
| 10933075 | |
| ≥98.0% (GC,T) | |
| 2-[4-(Trifluoromethyl)phenyl]pyridine |
| 77°C | |
| C12H8F3N | |
| 1 g | |
| FC(F)(F)C1=CC=C(C=C1)C1=CC=CC=N1 | |
| 223.20 | |
| 223.20 | |
| Crystalline Powder |
Safety and Handling
TSCA : No