accessibility menu, dialog, popup

UserName

CAS RN 203065-88-7

NFFF

CAS RN 203065-88-7

IUPAC Name: 2-[4-(trifluoromethyl)phenyl]pyridine
Synonyms: 2-[4-(trifluoromethyl)phenyl]pyridine
Molecular Weight (g/mol): 223.2
Molecular Formula: C12H8F3N
InChi Key: KGZSSIFFYUBVOX-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)C1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results