missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[4-(Trifluoromethyl)phenyl]pyridine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 77°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 223.20 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-[4-(Trifluoromethyl)phenyl]pyridine |
Safety and Handling
| TSCA | No |