missing translation for 'onlineSavingsMsg'
Learn More

2-[4-(Trifluoromethyl)phenyl]ethylamine, 98%

Catalog No. AAH6441503
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6441503 1 g
AAH64415MD 250 mg
AAH6441506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6441503 Supplier Thermo Scientific Chemicals Supplier No. H6441503

CAS: 775-00-8 | C9H10F3N | 189.181 g/mol

It is applied as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 775-00-8
Molecular Formula C9H10F3N
Molecular Weight (g/mol) 189.181
MDL Number MFCD01529875
InChI Key HMFOBPNVAAAACP-UHFFFAOYSA-N
Synonym 4-trifluoromethylphenethylamine, 2-4-trifluoromethyl-phenyl-ethylamine, 2-4-trifluoromethyl phenyl ethanamine, 4-trifluoromethyl phenethylamine, 2-4-trifluoromethyl-phenyl ethylamine, 2-4-trifluoromethylphenyl ethylamine, 2-4-trifluoromethyl phenyl ethan-1-amine, 2-4-trifluoromethyl phenyl ethylamine, 2-4-trifluoromethyl phenyl ethyl amine
PubChem CID 1988106
IUPAC Name 2-[4-(trifluoromethyl)phenyl]ethanamine
SMILES C1=CC(=CC=C1CCN)C(F)(F)F

Specifications

Quantity 1 g
UN Number UN2735
Beilstein 2719818
Solubility Information Slightly soluble in water.
Formula Weight 189.18
Percent Purity 98%
Chemical Name or Material 2-[4-(Trifluoromethyl)phenyl]ethylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only