missing translation for 'onlineSavingsMsg'
Learn More

2-(4-tert-Butylphenyl)pyridine 97.0+%, TCI America™

Catalog No. B4092200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4092200MG 200 mg
B40921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4092200MG Supplier TCI America Supplier No. B4092200MG
Only null left

Chemical Identifiers

CAS 524713-66-4
Molecular Formula C15H17N
Molecular Weight (g/mol) 211.31
MDL Number MFCD07775634
InChI Key RHXQSMVSBYAGQV-UHFFFAOYSA-N
PubChem CID 12984537
IUPAC Name 2-(4-tert-butylphenyl)pyridine
SMILES CC(C)(C)C1=CC=C(C=C1)C1=CC=CC=N1

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 211.31
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(4-tert-Butylphenyl)pyridine